2,5-Difluoro-4-formylphenylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde

33171-500MG 33171-1G 33171-5G 33171-25G
77325-472EA 155.53 USD
77325-472 77345-584 77361-830 77384-910
2,5-Difluoro-4-formylphenylphenylboronic acid pinacol ester ≥97%
2,5-Difluoro-4-formylphenylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BF₂O₃
MW: 268.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723176
CAS Number: 2121512-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR