[2,3'-Bipyridin]-5'-amine ≥95%

Supplier: AOBChem USA

Warning

33157-500MG 33157-1G 33157-5G 33157-25G
77325-468EA 284.28 USD
77325-468 77349-190 77361-826 77384-906
[2,3'-Bipyridin]-5'-amine ≥95%
[2,3'-Bipyridin]-5'-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MW: 171.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19544015
CAS Number: 1245745-55-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR