2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
Supplier: AOBChem USA
|
Warning
|
33152-500MG
33152-1G
33152-5G
33152-25G
77325-466EA
95.79
USD
77325-466
77342-604
77361-824
77373-442
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97%
2-(3,4-Dichloro-2-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₃H₁₇BCl₂O₃ MW: 302.99 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22415026 CAS Number: 2121513-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...