1-(3-Fluoro-(5-methylsulfanyl)phenyl)pyrolidine ≥97%

Supplier: AOBChem USA

Warning

33144-500MG 33144-1G 33144-5G 33144-25G
77325-460EA 140.08 USD
77325-460 77344-738 77361-818 77384-902
1-(3-Fluoro-(5-methylsulfanyl)phenyl)pyrolidine ≥97%
1-(3-Fluoro-(5-methylsulfanyl)phenyl)pyrolidine

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNS
Storage Temperature: Ambient
MDL Number: MFCD30723172
CAS Number: 2121515-14-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR