2-Fluoro-4-methylpyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33138-500MG 33138-1G 33138-5G
77325-458EA 314.15 USD
77325-458 77349-188 77373-438
2-Fluoro-4-methylpyridine-3-boronic acid pinacol ester ≥97%
2-Fluoro-4-methylpyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₇BFNO₂
Storage Temperature: Ambient
MDL Number: MFCD16995669
CAS Number: 2824977-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR