Ethyl 2-methoxypyrimidine-5-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33137-500MG 33137-1G 33137-5G 33137-25G
77325-456EA 243.08 USD
77325-456 77349-186 77361-816 77384-900
Ethyl 2-methoxypyrimidine-5-carboxylate ≥97%
Ethyl 2-methoxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₃
MW: 182.177 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18910361
CAS Number: 1378331-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR