2,5-Dimethoxy-4-methylphenol ≥95%

Supplier: AOBChem USA

Warning

33136-500MG 33136-1G 33136-5G
77325-454EA 338.87 USD
77325-454 77349-184 77373-436
2,5-Dimethoxy-4-methylphenol ≥95%
2,5-Dimethoxy-4-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00829468
CAS Number: 73120-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR