3-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(3-(Benzyloxy)-4-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Benzyloxy)-4-fluorophenylboronic acid pinacolester, 3-Benzyloxy-4-fluorobenzeneboronic acid pinacol ester

33098-500MG 33098-1G 33098-25G 33098-5G
77325-452EA 290.46 USD
77325-452 77349-180 77384-898 77373-432
3-Benzyloxy-4-fluorophenylboronic acid pinacol ester ≥97%
3-Benzyloxy-4-fluorophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₉H₂₂BFO₃
Storage Temperature: Ambient
MDL Number: MFCD23098800
CAS Number: 2121515-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR