N-(3-acetylpyridin-2-yl)pivalamide ≥97%

Supplier: AOBChem USA

Warning

33075-500MG 33075-1G 33075-5G 33075-25G
77325-450EA 143.17 USD
77325-450 77344-982 77361-814 77384-896
N-(3-acetylpyridin-2-yl)pivalamide ≥97%
N-(3-acetylpyridin-2-yl)pivalamide

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₂
MW: 220.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15526877
CAS Number: 161987-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR