5-(Methoxymethyl)-pyridine-3-carboxaldehyde ≥95%

Supplier: AOBChem USA

Warning

33044-500MG 33044-1G 33044-5G
77339-310EA 576.8 USD
77339-310 77349-178 77373-430
5-(Methoxymethyl)-pyridine-3-carboxaldehyde ≥95%
5-(Methoxymethyl)-pyridine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32707001
CAS Number: 1669438-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR