3,4-Dichloro-2-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

33026-500MG 33026-1G 33026-5G
77337-840EA 449.08 USD
77337-840 77349-176 77373-424
3,4-Dichloro-2-methoxybenzyl alcohol ≥97%
3,4-Dichloro-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD29762034
CAS Number: 1935620-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR