(3-Bromo-2-ethoxyphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

32963-500MG 32963-1G 32963-5G 32963-25G
77325-444EA 233.81 USD
77325-444 77349-172 77361-806 77384-892
(3-Bromo-2-ethoxyphenyl)methanol ≥97%
(3-Bromo-2-ethoxyphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD14695227
CAS Number: 1303899-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR