5-Bromo-3-(4-Bromophenylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32910-500MG 32910-1G 32910-5G 32910-25G
77325-438EA 208.06 USD
77325-438 77349-168 77361-800 77384-890
5-Bromo-3-(4-Bromophenylcarbamoyl)phenylboronic acid ≥97%
5-Bromo-3-(4-Bromophenylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₀BBr₂NO₃
Storage Temperature: Ambient
MDL Number: MFCD30529876
CAS Number: 2121515-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR