5-Bromo-3-(morpholine-4-carbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32907-500MG 32907-1G 32907-5G 32907-25G
77325-434EA 156.56 USD
77325-434 77345-606 77361-796 77384-886
5-Bromo-3-(morpholine-4-carbonyl)phenylboronic acid ≥97%
5-Bromo-3-(morpholine-4-carbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃BBrNO₄
MW: 313.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529874
CAS Number: 2121512-75-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR