5-Bromo-3-(piperidine-1-carbonyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
32906-500MG
32906-1G
32906-5G
32906-25G
77325-432EA
182.31
USD
77325-432
77346-910
77361-794
77384-884
5-Bromo-3-(piperidine-1-carbonyl)phenylboronic acid ≥97%
5-Bromo-3-(piperidine-1-carbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BBrNO₃ MW: 311.967 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28399069 CAS Number: 2121514-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...