5-Bromo-3-(pyrrolidine-1-carbonyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
32905-500MG
32905-1G
32905-5G
32905-25G
77325-430EA
134.93
USD
77325-430
77344-442
77361-792
77384-882
5-Bromo-3-(pyrrolidine-1-carbonyl)phenylboronic acid ≥97%
5-Bromo-3-(pyrrolidine-1-carbonyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₃BBrNO₃ MW: 297.941 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30529873 CAS Number: 2121514-36-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...