5-Bromo-3-(N,O-dimethylhydroxylaminocarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

32903-500MG 32903-1G 32903-5G 32903-25G
77325-426EA 134.93 USD
77325-426 77344-440 77361-788 77384-880
5-Bromo-3-(N,O-dimethylhydroxylaminocarbonyl)phenylboronic acid ≥97%
5-Bromo-3-(N,O-dimethylhydroxylaminocarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BBrNO₄
MW: 287.903 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529872
CAS Number: 2121514-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR