Methyl 3-chloro-2-methoxy-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32891-500MG 32891-1G 32891-5G 32891-25G
77325-420EA 140.08 USD
77325-420 77344-702 77361-782 77384-874
Methyl 3-chloro-2-methoxy-4-methylbenzoate ≥97%
Methyl 3-chloro-2-methoxy-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529866
CAS Number: 1936018-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR