[3-Fluoro-5-(pyrrolidin-1-yl)phenyl]methanol ≥95%

Supplier: AOBChem USA

Warning

32880-500MG 32880-1G 32880-5G 32880-25G
77325-418EA 191.58 USD
77325-418 77349-160 77361-780 77384-872
[3-Fluoro-5-(pyrrolidin-1-yl)phenyl]methanol ≥95%
[3-Fluoro-5-(pyrrolidin-1-yl)phenyl]methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄FNO
MW: 195.233 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805955
CAS Number: 1690605-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR