3-Chloro-2-ethylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

32850-500MG 32850-1G 32850-5G 32850-25G
77325-416EA 167.89 USD
77325-416 77346-222 77361-778 77384-870
3-Chloro-2-ethylbenzoic acid ≥95%
3-Chloro-2-ethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD18410965
CAS Number: 67648-15-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR