(4-Bromo-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
Supplier: AOBChem USA
|
Warning
|
32831-500MG
32831-1G
32831-5G
32831-25G
77325-414EA
163.77
USD
77325-414
77346-106
77361-776
77384-868
(4-Bromo-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone ≥97%
(4-Bromo-2-fluorophenyl)(4-methylpiperazin-1-yl)methanone
Precaución: for Research Use Only
Formula:
C₁₂H₁₄BrFN₂O MW: 301.155 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09942268 CAS Number: 924642-60-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...