5-Bromo-1-(cyclopropylmethyl)-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

32826-500MG 32826-1G 32826-5G 32826-25G
77325-412EA 182.31 USD
77325-412 77346-908 77361-774 77384-866
5-Bromo-1-(cyclopropylmethyl)-1H-indazole ≥97%
5-Bromo-1-(cyclopropylmethyl)-1H-indazole

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂
Storage Temperature: Ambient
MDL Number: MFCD28663073
CAS Number: 756839-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR