6-Bromo-3-iodo-1-methyl-1H-indazole ≥97%
Supplier: AOBChem USA
|
Warning
|
32823-500MG
32823-1G
32823-5G
32823-25G
77325-410EA
256.47
USD
77325-410
77349-158
77361-772
77384-864
6-Bromo-3-iodo-1-methyl-1H-indazole ≥97%
6-Bromo-3-iodo-1-methyl-1H-indazole
Precaución: for Research Use Only
Formula:
C₈H₆BrIN₂ Storage Temperature: Ambient |
MDL Number:
MFCD12756577 CAS Number: 1260741-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...