Methyl 3-chloro-5-fluoro-4-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32805-500MG 32805-1G 32805-5G
77337-638EA 432.6 USD
77337-638 77349-154 77373-404
Methyl 3-chloro-5-fluoro-4-hydroxybenzoate ≥97%
Methyl 3-chloro-5-fluoro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₃
MW: 204.583 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24718242
CAS Number: 369-15-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR