2-(Benzyloxy)-4-bromo-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32801-1G 32801-5G 32801-25G
77341-988EA 111.24 USD
77341-988 77361-768 77373-402
2-(Benzyloxy)-4-bromo-5-methylbenzaldehyde ≥97%
2-(Benzyloxy)-4-bromo-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD30529862
CAS Number: 2056110-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR