2-Chloro-5-(trifluoromethyl)-4-pyridinemethanol ≥97%

Supplier: AOBChem USA

Warning

32791-500MG 32791-1G 32791-5G 32791-25G
77325-404EA 263.68 USD
77325-404 77349-152 77361-762 77384-862
2-Chloro-5-(trifluoromethyl)-4-pyridinemethanol ≥97%
2-Chloro-5-(trifluoromethyl)-4-pyridinemethanol

Precaución: for Research Use Only

Formula: C₇H₅ClF₃NO
MW: 211.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16607489
CAS Number: 1227585-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR