5-(Benzyloxy)-2-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

32788-500MG 32788-1G 32788-5G
77325-402EA 341.96 USD
77325-402 77349-150 77373-396
5-(Benzyloxy)-2-fluoro-4-iodopyridine ≥97%
5-(Benzyloxy)-2-fluoro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₁₂H₉FINO
MW: 329.109 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529858
CAS Number: 2056110-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR