2-Fluoro-4-iodo-5-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

32787-500MG 32787-1G 32787-5G
77338-120EA 466.59 USD
77338-120 77349-148 77373-394
2-Fluoro-4-iodo-5-isopropoxypyridine ≥97%
2-Fluoro-4-iodo-5-isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₉FINO
Storage Temperature: Ambient
MDL Number: MFCD30529857
CAS Number: 2056110-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR