4-Benzyloxy-5-fluoro-3-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32778-500MG 32778-1G 32778-5G 32778-25G
77325-400EA 143.17 USD
77325-400 77344-998 77361-760 77384-860
4-Benzyloxy-5-fluoro-3-methoxybenzyl alcohol ≥97%
4-Benzyloxy-5-fluoro-3-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₁₅H₁₅FO₃
Storage Temperature: Ambient
MDL Number: MFCD30529856
CAS Number: 2056110-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR