3-Methyl-2-trifluoromethylpyridine-5-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32771-500MG 32771-1G 32771-5G
77338-238EA 470.71 USD
77338-238 77349-144 77373-390
3-Methyl-2-trifluoromethylpyridine-5-boronic acid pinacol ester ≥97%
3-Methyl-2-trifluoromethylpyridine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₃NO₂
MW: 287.086 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18726736
CAS Number: 2121511-95-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR