3-Chloro-2-methoxy-4-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32770-500MG 32770-1G 32770-5G 32770-25G
77325-398EA 122.57 USD
77325-398 77343-978 77361-758 77373-388
3-Chloro-2-methoxy-4-methylbenzyl alcohol ≥97%
3-Chloro-2-methoxy-4-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529854
CAS Number: 1784905-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR