3-Chloro-2-methoxy-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32769-500MG 32769-1G 32769-5G 32769-25G
77325-396EA 108.15 USD
77325-396 77343-158 77361-756 77373-386
3-Chloro-2-methoxy-4-methylbenzaldehyde ≥97%
3-Chloro-2-methoxy-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30529853
CAS Number: 1782217-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR