2-(Benzyloxy)-5-bromo-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32739-500MG 32739-1G 32739-5G 32739-25G
77325-392EA 199.82 USD
77325-392 77349-140 77361-752 77384-856
2-(Benzyloxy)-5-bromo-3-methylbenzaldehyde ≥97%
2-(Benzyloxy)-5-bromo-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD26059137
CAS Number: 1566412-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR