5-Bromo-2-ethoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32737-500MG 32737-1G 32737-5G 32737-25G
77325-390EA 196.73 USD
77325-390 77349-138 77361-750 77384-854
5-Bromo-2-ethoxy-3-methylbenzaldehyde ≥97%
5-Bromo-2-ethoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25342423
CAS Number: 708984-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR