3-Formyl-4-methoxy-5-methylphenyl boronic acid pinacol ester ≥95%
Supplier: AOBChem USA
|
Warning
|
32735-500MG
32735-1G
32735-5G
77337-838EA
449.08
USD
77337-838
77349-136
77373-384
3-Formyl-4-methoxy-5-methylphenyl boronic acid pinacol ester ≥95%
3-Formyl-4-methoxy-5-methylphenyl boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₁BO₄ Storage Temperature: Ambient |
MDL Number:
MFCD30529852 CAS Number: 2121511-84-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...