3-Formyl-4-methoxy-5-methylphenyl boronic acid ≥95%

Supplier: AOBChem USA

Warning

32734-500MG 32734-1G 32734-5G
77338-884EA 528.39 USD
77338-884 77349-134 77373-382
3-Formyl-4-methoxy-5-methylphenyl boronic acid ≥95%
3-Formyl-4-methoxy-5-methylphenyl boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MW: 193.992 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30723164
CAS Number: 2121512-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR