2-(Benzyloxy)-5-bromo-4-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32725-500MG 32725-1G 32725-5G 32725-25G
77325-388EA 222.48 USD
77325-388 77349-132 77361-748 77384-852
2-(Benzyloxy)-5-bromo-4-methylbenzaldehyde ≥97%
2-(Benzyloxy)-5-bromo-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₂
MW: 305.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25459888
CAS Number: 1823782-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR