(5-Chloro-2-ethoxypyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

32682-500MG 32682-1G 32682-5G 32682-25G
77325-378EA 243.08 USD
77325-378 77349-120 77361-740 77384-844
(5-Chloro-2-ethoxypyridin-4-yl)methanol ≥97%
(5-Chloro-2-ethoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529847
CAS Number: 1595902-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR