5-Chloro-2-ethoxypyridine-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

32679-500MG 32679-1G 32679-5G
77325-376EA 346.08 USD
77325-376 77349-116 77373-374
5-Chloro-2-ethoxypyridine-4-boronic acid pinacol ester ≥97%
5-Chloro-2-ethoxypyridine-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BClNO₃
MW: 283.559 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529845
CAS Number: 2121513-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR