3-Fluoroisonicotinaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

32656-500MG 32656-1G 32656-5G 32656-25G
77325-374EA 154.5 USD
77325-374 77345-550 77361-738 77384-842
3-Fluoroisonicotinaldehyde oxime ≥97%
3-Fluoroisonicotinaldehyde oxime

Precaución: for Research Use Only

Formula: C₆H₅FN₂O
MW: 140.115 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28384887
CAS Number: 471909-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR