4-(Benzyloxy)-3-chloro-5-fluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

32652-500MG 32652-1G 32652-5G
77335-954EA 370.8 USD
77335-954 77349-112 77373-370
4-(Benzyloxy)-3-chloro-5-fluorobenzaldehyde ≥95%
4-(Benzyloxy)-3-chloro-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD30529840
CAS Number: 1709785-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR