3-Chloro-2,4-dimethylbenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

32637-500MG 32637-1G 32637-5G 32637-25G
77325-372EA 259.56 USD
77325-372 77349-110 77361-736 77384-840
3-Chloro-2,4-dimethylbenzoic acid methyl ester ≥97%
3-Chloro-2,4-dimethylbenzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30529835
CAS Number: 2056110-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR