3-Chloro-2,4-dimethylbenzyl alcohol ≥95%

Supplier: AOBChem USA

Warning

32636-500MG 32636-1G 32636-5G 32636-25G
77325-370EA 243.08 USD
77325-370 77349-108 77361-734 77384-838
3-Chloro-2,4-dimethylbenzyl alcohol ≥95%
3-Chloro-2,4-dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD30529834
CAS Number: 1780815-77-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR