4-(4,4-Dimethylcyclohexyl)-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

32624-500MG 32624-1G 32624-25G 32624-5G
77325-368EA 289.43 USD
77325-368 77349-106 77384-836 77373-368
4-(4,4-Dimethylcyclohexyl)-benzoic acid ≥97%
4-(4,4-Dimethylcyclohexyl)-benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₂
MW: 232.318 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34755914
CAS Number: 1784095-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR