3-Chloro-2-ethoxypyridine-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

32613-500MG 32613-1G 32613-5G 32613-25G
77325-364EA 249.26 USD
77325-364 77349-100 77361-732 77384-834
3-Chloro-2-ethoxypyridine-4-carboxylic acid ≥97%
3-Chloro-2-ethoxypyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529829
CAS Number: 1696892-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR