(3-Chloro-2-ethoxypyridin-4-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

32612-500MG 32612-1G 32612-5G 32612-25G
77325-362EA 263.68 USD
77325-362 77349-098 77361-730 77384-832
(3-Chloro-2-ethoxypyridin-4-yl)methanol ≥95%
(3-Chloro-2-ethoxypyridin-4-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₀ClNO₂
MW: 187.624 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30529828
CAS Number: 2056110-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR