2-(Benzyloxy)-5-bromo-1-chloro-3-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

32604-500MG 32604-1G 32604-5G 32604-25G
77325-360EA 183.34 USD
77325-360 77347-114 77361-728 77384-830
2-(Benzyloxy)-5-bromo-1-chloro-3-fluorobenzene ≥97%
2-(Benzyloxy)-5-bromo-1-chloro-3-fluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrClFO
MW: 315.565 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29043957
CAS Number: 1881293-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR