2-Benzyloxy-5-methoxy-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32555-1G 32555-5G 32555-25G
77340-890EA 62.83 USD
77340-890 77361-724 77373-358
2-Benzyloxy-5-methoxy-benzaldehyde ≥97%
2-Benzyloxy-5-methoxy-benzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00436294
CAS Number: 56979-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR