(2-Ethoxy-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

32549-500MG 32549-1G 32549-5G 32549-25G
77325-356EA 99.91 USD
77325-356 77342-770 77361-722 77373-356
(2-Ethoxy-5-methylphenyl)methanol ≥97%
(2-Ethoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
Storage Temperature: Ambient
MDL Number: MFCD18331419
CAS Number: 1409767-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR