Benzyl 3-chloro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32538-1G 32538-5G 32538-25G
77341-406EA 85.49 USD
77341-406 77361-720 77373-354
Benzyl 3-chloro-4-methylbenzoate ≥97%
Benzyl 3-chloro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₂
Storage Temperature: Ambient
MDL Number: MFCD30529822
CAS Number: 2056110-37-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR